Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0140

Sigma-Aldrich

Magnesium carbonate basic

SAJ special grade, 40.0-45.0% MgO basis

Synonym(s):

Magnesium hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCO3
CAS Number:
Molecular Weight:
84.31
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ special grade
Pricing and availability is not currently available.

grade

SAJ special grade

availability

available only in Japan

concentration

40.0-45.0% (MgO)

dilution

(for analytical testing)

SMILES string

[Mg++].O[Mg]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Mg.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

UDYRPJABTMRVCX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service