Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0090

Sigma-Aldrich

Magnesium acetate tetrahydrate

SAJ special grade, ≥99.0%

Synonym(s):

Acetic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45
Beilstein/REAXYS Number:
3730605
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ special grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥99.0%

form

solid

availability

available only in Japan

mp

72-75 °C (lit.)

SMILES string

O.O.O.O.CC(=O)O[Mg]OC(C)=O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihito Kitaoku et al.
Planta, 242(4), 895-907 (2015-05-23)
We first solved the crystal structure of class III catalytic domain of a chitinase from fern (PrChiA-cat), and found a structural difference between PrChiA-cat and hevamine. PrChiA-cat was found to have reduced affinities to chitin oligosaccharides and allosamidin. Plant class
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the
Andrey G Malyutin et al.
ACS applied materials & interfaces, 7(22), 12089-12098 (2015-05-20)
Here we explore the formation of virus-like nanoparticles (VNPs) utilizing 22-24 nm iron oxide nanoparticles (NPs) as cores and proteins derived from viral capsids of brome mosaic virus (BMV) or hepatitis B virus (HBV) as shells. To accomplish that, hydrophobic
Tanja Rozman Peterka et al.
Journal of pharmaceutical and biomedical analysis, 110, 67-75 (2015-03-26)
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service