Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18-0830

Sigma-Aldrich

Lead oxide, red

SAJ first grade, ≥95.0%

Synonym(s):

Lead(II,IV) oxide, Minium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb3O4
CAS Number:
Molecular Weight:
685.60
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥95.0%
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

10 mmHg ( 0 °C)

assay

≥95.0%

form

solid

availability

available only in Japan

SMILES string

O1[PbH2]O[Pb]12O[PbH2]O2

InChI

1S/4O.3Pb

InChI key

XMFOQHDPRMAJNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Ox. Sol. 2 - Repr. 1A - STOT RE 1

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujun Zhang et al.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control, 52(11), 2131-2139 (2006-01-21)
The perovskite solid solution system (1-x)BiScO3-(x)PbTiO3 represents an interesting new family of high-temperature piezoelectric materials. Compositions near the morphotropic phase boundary (x approximately 0.64) have been reported to have high Curie temperatures (Tc > 450 degrees C) and good piezoelectric
S P Gabuda et al.
Solid state nuclear magnetic resonance, 15(2), 103-107 (2000-02-12)
Solid Pb3O4 has been studied with 207Pb nuclear magnetic resonance (NMR) spectroscopy. The 207Pb NMR chemical-shift tensor of the Pb2+ site has principal values of delta11 = 1980 +/- 5 ppm, delta22 = 1540 +/- 5 ppm, and delta33 =
W Thiel
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft, 174(3), 197-200 (1992-06-01)
Masses for arterial injection were evaluated in 360 whole cadavers that had been injected with preservation fluid. A mass consisting of dextrin, latex, and lead tetroxide produced the best results. This mass provided high radiopacity at large distances from the
[Abnormal lead absorption due to penetration of lead tetraoxide (red-lead) in the hand (author's transl)].
F Loi
La Medicina del lavoro, 71(2), 157-160 (1980-03-01)
T Kákosy et al.
Journal of toxicology. Clinical toxicology, 34(5), 507-511 (1996-01-01)
Report of a lead intoxication epidemic caused by ingestion of contaminated ground paprika. Retrospective study of case histories. Institutional, toxicological and medical referral center for ambulatory and hospitalized care. One hundred forty-one adults consumed paprika contaminated with lead tetroxide (red

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service