Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16-0040

Sigma-Aldrich

Japanese acid clay

CP

Synonym(s):

Acid clay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
grade:
CP

grade

CP

availability

available only in Japan

dilution

(for analytical testing)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service