Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-2461

Sigma-Aldrich

Isopropyl β-D-1-thiogalactopyranoside

≥99.0%

Synonym(s):

IPTG, Isopropyl β-D-thiogalactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
Beilstein/REAXYS Number:
4631
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥99.0%

form

solid

availability

available only in Japan

SMILES string

CC(C)S[C@@H]1OC(CO)[C@H](O)C(O)[C@H]1O

InChI

1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1

InChI key

BPHPUYQFMNQIOC-NXRLNHOXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

IPTG is commonly used in cloning procedures that require induction of β-galactosidase activity. It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.

Other Notes

Non-metabolizable galactose analog.

Suitability

for biological purposes

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therese Wohlschlager et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), E2787-E2796 (2014-06-01)
Effector proteins of innate immune systems recognize specific non-self epitopes. Tectonins are a family of β-propeller lectins conserved from bacteria to mammals that have been shown to bind bacterial lipopolysaccharide (LPS). We present experimental evidence that two Tectonins of fungal
Simon van Vliet et al.
BMC microbiology, 14, 116-116 (2014-06-03)
Bacterial habitats, such as soil and the gut, are structured at the micrometer scale. Important aspects of microbial life in such spatial ecosystems are migration and colonization. Here we explore the colonization of a structured ecosystem by two neutrally labeled
Iltaf Ahmed et al.
Human molecular genetics, 24(11), 3172-3180 (2015-02-24)
There are two known mRNA degradation pathways, 3' to 5' and 5' to 3'. We identified likely pathogenic variants in two genes involved in these two pathways in individuals with intellectual disability. In a large family with multiple branches, we
Farshid Hadifar et al.
PloS one, 9(10), e108420-e108420 (2014-10-21)
Killed avian influenza virus (AIV) vaccines have been used to control H5N1 infections in countries where the virus is endemic. Distinguishing vaccinated from naturally infected birds (DIVA) in such situations however, has become a major challenge. Recently, we introduced the
A P Carneiro et al.
British journal of cancer, 111(3), 551-558 (2014-06-18)
This study aimed to identify novel biomarkers for thyroid carcinoma diagnosis and prognosis. We have constructed a human single-chain variable fragment (scFv) antibody library that was selected against tumour thyroid cells using the BRASIL method (biopanning and rapid analysis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service