Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15-1310

Sigma-Aldrich

Iron(III) nitrate nonahydrate

SAJ first grade, ≥98.0%

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98.0%
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥98.0%

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

1.5 (20 °C)

mp

47 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Runjuan Wang et al.
Bioresource technology, 120, 290-294 (2012-07-10)
The study investigated the effects of two-step pretreatment on fermentable sugar production from corn stalk silage. In the first step, the corn stalk silage was extracted by tepid water and then the solid was pretreated using Fe(NO(3))(3) as catalyst. The
Mara Ghiazza et al.
Chemical research in toxicology, 24(1), 99-110 (2010-12-22)
The mechanism of enhancement/inhibition of quartz toxicity induced by iron is still unclear. Here the amount of iron on a fibrogenic quartz (Qz) was increased by wet impregnation (Fe(NO(3))(3) 0.67 and 6.7 wt %). X-ray diffraction (XRD), XRF diffuse reflectance
F Ndong et al.
International endodontic journal, 45(6), 557-564 (2012-01-25)
To determine the effect of increasing the proportion of zinc on partially stabilized cement (PSC) produced using a one-step sol gel process. A one-step sol-gel process of Portland cement-based PSC with Zn was synthesized by replacing iron nitrate. The crystalline
Marek Basler et al.
Proteomics, 6(23), 6194-6206 (2006-11-30)
Restricting bacterial growth by iron-chelating proteins that reduce iron availability in mucosal secretions and body fluids belongs to basic mechanisms of innate immunity. Most pathogens and commensals thus developed gene regulons responding to iron concentration and encoding iron acquisition systems
Xianmao Lu et al.
Nano letters, 7(6), 1764-1769 (2007-05-11)
This paper describes a two-step procedure for generating cubic nanocages and nanoframes. In the first step, Au/Ag alloy nanoboxes were synthesized through the galvanic replacement reaction between Ag nanocubes and an aqueous HAuCl4 solution. The second step involved the selective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service