Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13-1610

Sigma-Aldrich

Hydrobromic acid

SAJ first grade, 45.0-49.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
CAS Number:
Molecular Weight:
80.91
Beilstein/REAXYS Number:
3587158
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

2.8 (vs air)

vapor pressure

320 psi ( 21.1 °C)
8 mmHg ( 25 °C)

description

48% aqueous HBr

form

liquid

availability

available only in Japan

concentration

45.0-49.0%

density

1.49 g/mL at 25 °C (lit.)

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujing Liang et al.
Theranostics, 5(9), 970-984 (2015-07-15)
Developing safe and effective nanoprobes for targeted imaging and selective therapy of gastric cancer stem cells (GCSCs) has become one of the most promising anticancer strategies. Herein, gold nanostars-based PEGylated multifunctional nanoprobes were prepared with conjugated CD44v6 monoclonal antibodies (CD44v6-GNS)
Po-Yu Tsai et al.
Physical chemistry chemical physics : PCCP, 13(4), 1419-1423 (2010-11-27)
The orientation dependence of Br-atom formation in the reaction of the oriented OH radical with the HBr molecule using the hexapole electrostatic field was studied. Experimental results for the orientation dependence in the reaction were analyzed using a Legendre polynomial
Ahmed Hasbi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(11), 4806-4820 (2014-07-27)
Although the dopamine D1-D2 receptor heteromer has emerging physiological relevance and a postulated role in different neuropsychiatric disorders, such as drug addiction, depression, and schizophrenia, there is a need for pharmacological tools that selectively target such receptor complexes in order
Arnaldur Hall et al.
Biomacromolecules, 16(7), 2119-2126 (2015-06-09)
Poly(L-lysine)s (PLLs), and related derivatives, have received considerable attention as nonviral vectors. High molecular weight PLLs (H-PLLs) are superior transfectants compared with low Mw PLLs (L-PLLs), but suggested to be more cytotoxic. Through a pan-integrated metabolomic approach using Seahorse XF
Małgorzata Olejniczak et al.
The Journal of chemical physics, 136(1), 014108-014108 (2012-01-14)
We report the implementation of nuclear magnetic resonance (NMR) shielding tensors within the four-component relativistic Kohn-Sham density functional theory including non-collinear spin magnetization and employing London atomic orbitals to ensure gauge origin independent results, together with a new and efficient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service