Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13-1510

Sigma-Aldrich

Hydrazine monohydrate

SAJ special grade, ≥98.0%

Synonym(s):

Hydrazinium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2O
CAS Number:
Molecular Weight:
50.06
Beilstein/REAXYS Number:
878137
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98.0%
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

>1 (vs air)

vapor pressure

5 mmHg ( 25 °C)

assay

≥98.0%

form

liquid

expl. lim.

99.99 %

availability

available only in Japan

refractive index

n20/D 1.428 (lit.)

pH

10.6-10.7 (10 g/L)

bp

120.1 °C (lit.)

mp

−51.7 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

O.NN

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2

InChI key

IKDUDTNKRLTJSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

167.0 °F

flash_point_c

75 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boran Kartal et al.
FEMS microbiology reviews, 37(3), 428-461 (2012-12-06)
Anaerobic ammonium-oxidizing (anammox) bacteria primarily grow by the oxidation of ammonium coupled to nitrite reduction, using CO2 as the sole carbon source. Although they were neglected for a long time, anammox bacteria are encountered in an enormous species (micro)diversity in
Yinhu Wang et al.
Biosensors & bioelectronics, 43, 180-185 (2013-01-12)
We report a novel hydrogen peroxide (H2O2) and hydrazine sensor based on low-cost poly(vinylpyrrolidone)-protected silver nanocubes (PVP-AgNCs). The monodisperse silver nanocubes were prepared by adding a trace amount of sodium sulfide in the conventional polyol synthesis for fast reduction of
Tino Wilson Sanchez et al.
Bioorganic & medicinal chemistry, 21(4), 957-963 (2013-01-12)
Human lens epithelium-derived growth factor (LEDGF)/p75 plays an important role in the HIV life cycle by stimulating integrase (IN)-led viral DNA integration into cellular chromosomes. Mechanistic studies show the majority of IN inhibitors chelate magnesium ions in the catalytic active
Timothy M Alligrant et al.
Lab on a chip, 13(3), 349-354 (2012-12-06)
We report on real-time electrochemical detection of individual DNA hybridization events at an electrode surface. The experiment is carried out in a microelectrochemical device configured with a working electrode modified with single-stranded DNA probe molecules. When a complementary DNA strand
Mild and rapid Pd-catalyzed cross-coupling with hydrazine in continuous flow: application to the synthesis of functionalized heterocycles.
Andrew DeAngelis et al.
Angewandte Chemie (International ed. in English), 52(12), 3434-3437 (2013-02-14)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service