Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13-1190

Sigma-Aldrich

L-Histidine monohydrochloride monohydrate

SAJ special grade, ≥99.0%

Synonym(s):

L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein/REAXYS Number:
4168261
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ special grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ special grade

vapor pressure

<0.1 hPa ( 20 °C)

assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

4.38-4.48 (20 °C, 149.55 g/L)

mp

254 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Maldonado et al.
Neurochemistry international, 85-86, 31-39 (2015-05-06)
The effect of reserpine on histamine (HA) and tele-methylhistamine (N(τ)-MHA) in hypothalamus and cortex of rats was analyzed and compared to catecholamines. IP injection of reserpine (5 mg/kg) confirmed the effectiveness of reserpine treatment on noradrenaline and dopamine levels. Our in-vitro
Verena Saller et al.
Journal of pharmaceutical sciences, 104(4), 1440-1450 (2015-01-22)
In a typical manufacturing setup for biopharmaceutical drug products, the fill and dosing pump is placed after the final sterile filtration unit in order to ensure adequate dispensing accuracy and avoid backpressure peaks. Given the sensitivity of protein molecules, peristaltic
Aldo Roda et al.
The Analyst, 139(24), 6494-6501 (2014-10-25)
Increasingly, smartphones are used as portable personal computers, revolutionizing communication styles and entire lifestyles. Using 3D-printing technology we have made a disposable minicartridge that can be easily prototyped to turn any kind of smartphone or tablet into a portable luminometer
Kristin Bjornsdottir-Butler et al.
Journal of food protection, 78(7), 1335-1342 (2015-07-22)
Recent developments in detection and enumeration of histamine-producing bacteria (HPB) have created powerful molecular-based tools to better understand the presence of spoilage bacteria and conditions, resulting in increased risk of scombrotoxin fish poisoning. We examined 235 scombrotoxin-forming fish from the
Nam Ah Kim et al.
Drug development and industrial pharmacy, 41(2), 300-306 (2014-02-08)
Correlation of thermodynamic and secondary structural stability of proteins at various buffer pHs was investigated using differential scanning calorimetry (DSC), dynamic light scattering (DLS) and attenuated total reflection Fourier-transform infrared spectroscopy (ATR FT-IR). Recombinant human epithelial growth factor (rhEGF) was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service