Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-1840

Sigma-Aldrich

Tragacanth

CP

Synonym(s):

Gum tragacanth

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Koppang et al.
The Journal of prosthetic dentistry, 73(5), 486-491 (1995-05-01)
An in vitro test is described that simulates the in vivo fate of a denture adhesive, namely destruction, dilution, and dissolution of the adhesive, by repeated measurement of tensile bond strength for the adhesive in isotonic saline over time. The
Alteration in the tranquilizing potency of chlorpromazine in rats exposed chronically to the insecticide, endosulfan.
V Paul et al.
Bulletin of environmental contamination and toxicology, 53(5), 655-662 (1994-11-01)
Samira Alijani et al.
International journal of biological macromolecules, 49(4), 471-479 (2011-06-15)
In this study, Iranian gum tragacanth (GT) exudates from Astragalus fluccosus (AFG) and Astragalus gossypinus (AGG) were irradiated at 3, 7, 10 and 15 kGy. Fourier transform infrared spectroscopy (FTIR) data showed that irradiation did not induce changes in the
Zafar Iqbal et al.
Pakistan journal of pharmaceutical sciences, 24(4), 435-443 (2011-10-01)
Conventional dosage form is nowadays mostly replaced by sustained release formulation in order to increase drug efficacy and patient compliance. The sustained release properties of the PVP K90 alone and in combination with guar gum, xanthan gum and gum tragacanth
S Aziznia et al.
Journal of dairy science, 91(7), 2545-2552 (2008-06-21)
The effect of whey protein concentrate (WPC) and gum tragacanth (GT) as fat replacers on the chemical, physical, and microstructural properties of nonfat yogurt was investigated. The WPC (7.5, 15, and 20 g/L) and GT (0.25, 0.5, 0.75, and 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service