Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-0950

Sigma-Aldrich

L-Glutamine

JIS special grade, ≥99.0%

Synonym(s):

(S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Beilstein/REAXYS Number:
1723797
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

assay

≥99.0%

availability

available only in Japan

mp

185 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebekah M Samsonraj et al.
Stem cells (Dayton, Ohio), 33(6), 1878-1891 (2015-03-11)
This study sought to identify critical determinants of mesenchymal stem cell (MSC) potency using in vitro and in vivo attributes of cells isolated from the bone marrow of age- and sex-matched donors. Adherence to plastic was not indicative of potency
Lisa A Reynolds et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2984-2993 (2014-08-13)
Helminth parasites remain one of the most common causes of infections worldwide, yet little is still known about the immune signaling pathways that control their expulsion. C57BL/6 mice are chronically susceptible to infection with the gastrointestinal helminth parasite Heligmosomoides polygyrus.
Kohichi Matsunaga et al.
The Journal of cell biology, 190(4), 511-521 (2010-08-18)
Autophagy is a catabolic process that allows cells to digest their cytoplasmic constituents via autophagosome formation and lysosomal degradation. Recently, an autophagy-specific phosphatidylinositol 3-kinase (PI3-kinase) complex, consisting of hVps34, hVps15, Beclin-1, and Atg14L, has been identified in mammalian cells. Atg14L
Pelin Sahlén et al.
Genome biology, 16, 156-156 (2015-08-28)
Although the locations of promoters and enhancers have been identified in several cell types, we still have limited information on their connectivity. We developed HiCap, which combines a 4-cutter restriction enzyme Hi-C with sequence capture of promoter regions. Applying the
Ya-Chun Chen et al.
Nature genetics, 47(7), 803-808 (2015-05-26)
Pain perception has evolved as a warning mechanism to alert organisms to tissue damage and dangerous environments. In humans, however, undesirable, excessive or chronic pain is a common and major societal burden for which available medical treatments are currently suboptimal.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service