Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11-0753

Sigma-Aldrich

Formamide

≥99.5%

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein/REAXYS Number:
505995
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
assay:
≥99.5%
technique(s):
hybridization: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
Pricing and availability is not currently available.

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

assay

≥99.5%

form

liquid

autoignition temp.

932 °F

availability

available only in Japan

technique(s)

hybridization: suitable

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Suitability

for biological purposes

Preparation Note

deionized

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

target_organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

309.2 °F

flash_point_c

154 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Chen et al.
Prenatal diagnosis, 35(6), 534-543 (2015-02-03)
To develop a single-tube multi-marker assay for improved preimplantation genetic diagnosis (PGD) of deletional and/or non-deletional Hb Bart's hydrops fetalis syndrome, providing haplotype confirmation of deletional status, and maximization of linkage informativity. We performed in silico mining to identify novel
Jing Wang et al.
The journal of physical chemistry. B, 117(8), 2314-2320 (2013-01-26)
A step-by-step mechanistic pathway following the transformation of formamide to purine through a five-membered ring intermediate has been explored by density functional theory computations. The highlight of the mechanistic route detailed here is that the proposed pathway represents the simplest
Mado Vandewoestyne et al.
International journal of legal medicine, 123(5), 441-447 (2009-03-26)
Laser microdissection is a valuable tool for isolating specific cells from mixtures, such as male cells in a mixture with female cells, e.g., in cases of sexual assault. These cells can be stained with Y-chromosome-specific probes. We developed an automatic
Penelope A Bryant et al.
PloS one, 6(5), e19556-e19556 (2011-06-10)
A central issue in the design of microarray-based analysis of global gene expression is that variability resulting from experimental processes may obscure changes resulting from the effect being investigated. This study quantified the variability in gene expression at each level
Björn Heindryckx et al.
Mitochondrion, 18, 12-17 (2014-08-28)
To investigate the applicability of preimplantation genetic diagnosis (PGD), we used trophectoderm (TE) biopsy to determine the mutation load in a 35-year-old female with mitochondrial encephalopathy, lactic acidosis and stroke-like syndrome (MELAS). Transfer of a mutation-free blastocyst gave birth to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service