Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-0910

Sigma-Aldrich

Ethyl 4-aminobenzoate

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥99.0%

form

solid

availability

available only in Japan

mp

88-90 °C (lit.)

SMILES string

CCOC(=O)c1ccc(N)cc1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3

InChI key

BLFLLBZGZJTVJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

233.6 °F

flash_point_c

112 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathalie Ferreira Silva De Melo et al.
Journal of pharmaceutical sciences, 101(3), 1157-1165 (2011-11-23)
This paper describes a comparison of different polymeric nanocapsules (NCs) prepared with the polymers poly(D,L-lactide-co-glycolide), poly(L-lactide) (PLA), and poly(ε-caprolactone) and used as carrier systems for the local anesthetic (LA) benzocaine (BZC). The systems were characterized and their anesthetic activities investigated.
Qing Zhu et al.
Molecular pharmaceutics, 9(3), 546-553 (2012-02-04)
The crystallization kinetics of various active pharmaceutical ingredient/polyethylene glycol (API/PEG) solid dispersions has been investigated using wide-angle X-ray diffraction (XRD) and Raman spectroscopy. APIs with different physicochemical properties and crystallization tendency were employed to form solid dispersions with PEG. The
Sherry I Bame et al.
American journal of preventive medicine, 43(6 Suppl 5), S435-S442 (2012-11-28)
Hurricanes Katrina and Rita struck the Gulf Coast forcing unprecedented mass evacuation and devastation. Texas 2-1-1 is a disaster communication hub between callers with unmet needs and community services at disaster sites and evacuation destinations. To describe the location and
Sora Lee et al.
The Journal of general physiology, 139(6), 507-516 (2012-05-30)
Recent structural breakthroughs with the voltage-gated sodium channel from Arcobacter butzleri suggest that such bacterial channels may provide a structural platform to advance the understanding of eukaryotic sodium channel gating and pharmacology. We therefore set out to determine whether compounds
L A Linares et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 18(3), 267-269 (1990-07-01)
Methemoglobinemia is a condition in which more than 2% of the hemoglobin of the blood has been oxidized to the ferric form, a molecule which is incapable of binding oxygen or carbon dioxide. Clinically, the patient appears lethargic, cyanotic, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service