Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-1695

Sigma-Aldrich

Dibutyltin dilaurate

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2Sn[OCO(CH2)10CH3]2
CAS Number:
Molecular Weight:
631.56
Beilstein/REAXYS Number:
4156980
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

0.2 mmHg ( 160 °C)

form

solid

availability

available only in Japan

refractive index

n20/D 1.471 (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC(=O)O[Sn](CCCC)(CCCC)OC(=O)CCCCCCCCCCC

InChI

1S/2C12H24O2.2C4H9.Sn/c2*1-2-3-4-5-6-7-8-9-10-11-12(13)14;2*1-3-4-2;/h2*2-11H2,1H3,(H,13,14);2*1,3-4H2,2H3;/q;;;;+2/p-2

InChI key

UKLDJPRMSDWDSL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 1

target_organs

thymus, thymus,Immune system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

372.2 - 379.4 °F - closed cup

flash_point_c

189 - 193 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Subramoniam et al.
Toxicology letters, 57(3), 245-250 (1991-08-01)
Oral administration of a single dose of dibutyltin dilaurate (DBTL, 80 mg/kg body wt.), 2 or 24 h after treatment, caused no significant change in the levels of diacylglycerol and phosphoinositides in rat cerebrum (forebrain), whereas daily administration of DBTL
M A Khaliq et al.
Toxicology letters, 55(2), 179-183 (1991-02-01)
Administration of dibutyltin dilaurate (DBTL; 0, 20 or 40 mg/kg body wt.) by gavage to rats for 3 consecutive days produced a significant increase in polyamine levels in selected brain areas. At the higher dose of DBTL (40 mg/kg) spermidine
M Mushtaq et al.
Drug and chemical toxicology, 4(1), 75-88 (1981-01-01)
Toxicological studies of a leachable stabilizer Di-n-butyltin dilaurate (DBTL) were undertaken. Effects of DBTL after 15 days oral exposure to rats were studied on brain and liver enzyme activities. A significant decrease in body weight gain of DBTL exposed rats
Age and sex related behavioral changes induced by dibutyltin-dilaurate in rats.
M S Alam et al.
Bulletin of environmental contamination and toxicology, 50(2), 286-292 (1993-02-01)
P Chaynes et al.
Surgical and radiologic anatomy : SRA, 26(3), 235-238 (2004-04-21)
S3, S6 and S10 are the most commonly used agents for tissue plastination. Surprisingly, their chemical structures are not known. We therefore decided to fully characterize these products by standard analytical methods: multinucleus magnetic resonance, infrared spectroscopy and size exclusion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service