Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-0670

Sigma-Aldrich

Dextrin from potato starch

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H12O6)x
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshikazu Sugimoto et al.
Journal of industrial microbiology & biotechnology, 38(12), 1985-1991 (2011-05-28)
Submerged batch cultures of Aspergillus kawachii grown on indigestible dextrin were investigated for potential improvements in glucoamylase (GA) production. In flask culture, specific GA productivities per dry weight biomass using dextrin and indigestible dextrin were 11.0 and 56.1 mU/mg-DW, respectively. Indigestible
O E Odeleye et al.
European journal of clinical nutrition, 46(5), 367-371 (1992-05-01)
The effect of caloreen (glucose polymer) supplementation on indicators of iron status during protein-energy malnutrition was studied. Sixty-four children with moderate protein energy malnutrition (PEM) were fed diets supplemented with caloreen or starch (control) for 14 days, following which iron
Glucose polymer supplements in low birth weight infants.
O G Brooke
British medical journal (Clinical research ed.), 286(6371), 1143-1143 (1983-04-02)
C R Newton et al.
Journal of the Royal Society of Medicine, 78(1), 27-34 (1985-01-01)
The effectiveness of 5 different solutions on the absorption of fluid and electrolytes was tested in 7 patients with a proximal intestinal stoma and large fluid losses, all of whom previously needed intravenous infusions to maintain balance. In 4 patients
Glucose-polymer in acute diarrhoea.
Archives of disease in childhood, 57(9), 721-722 (1982-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service