Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-1820

Sigma-Aldrich

Carnauba wax

CP

Synonym(s):

Carnauba wax No. 1 yellow, Brazil wax

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

CP

form

chips

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reena Singh et al.
AAPS PharmSciTech, 8(3), E74-E74 (2007-10-06)
The purpose of the present study was to investigate incorporation of hydrophobic (ie, waxy) material into pellets using a thermal sintering technique and to evaluate the pellets in vitro for controlled release. Pellets prepared by extrusion-spheronization technology were formulated with
Sorada Kanokpanont et al.
International journal of pharmaceutics, 436(1-2), 141-153 (2012-07-10)
In this study, the novel silk fibroin-based bi-layered wound dressing was developed. Wax-coated silk fibroin woven fabric was introduced as a non-adhesive layer while the sponge made of sericin and glutaraldehyde-crosslinked silk fibroin/gelatin was fabricated as a bioactive layer. Wax-coated
Soheila Kheradmandnia et al.
Nanomedicine : nanotechnology, biology, and medicine, 6(6), 753-759 (2010-07-06)
Solid lipid nanoparticles (SLNs) have been proposed as suitable colloidal carriers for delivery of drugs with limited solubility. Ketoprofen as a model drug was incorporated into SLNs prepared from a mixture of beeswax and carnauba wax using Tween 80 and
William G Meikle et al.
Experimental & applied acarology, 46(1-4), 105-117 (2008-05-29)
Bee colonies in southern France were treated with conidia (asexual spores) from two strains of Beauveria bassiana, an entomopathogenic fungus. One strain was commercial (GHA) and the other had been isolated from Varroa mites in the region (Bb05002). Objectives were
Anna Giulia Balducci et al.
International journal of pharmaceutics, 421(2), 293-300 (2011-10-18)
The goal was to make available a delayed-release dosage form of mesalazine to be dispersed in water to facilitate swallowing in adults and children. Mesalazine microparticles containing carnauba wax were prepared by spray-congealing technique. A second step of spray-congealing of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service