Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-0870

Sigma-Aldrich

Calcium nitrate tetrahydrate

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
236.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥98.0%

form

solid

availability

available only in Japan

mp

44 °C (dec.) (lit.)

SMILES string

O.O.O.O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ca.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

ICSSIKVYVJQJND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Razavi et al.
Materials science & engineering. C, Materials for biological applications, 48, 21-27 (2015-01-13)
Although magnesium (Mg) is a unique biodegradable metal which possesses mechanical property similar to that of the natural bone and can be an attractive material to be used as orthopedic implants, its quick corrosion rate restricts its actual clinical applications.
Da-Young Seong et al.
Journal of photochemistry and photobiology. B, Biology, 146, 34-43 (2015-03-22)
Although methylene blue (MB) is the most inexpensive photosensitizer with promising applications in the photodynamic therapy (PDT) for its high quantum yield of singlet oxygen generation, the clinical use of MB has been limited by its rapid enzymatic reduction in
Senthilguru Kulanthaivel et al.
Materials science & engineering. C, Materials for biological applications, 58, 648-658 (2015-10-20)
The present study delineates the synthesis and characterization of cobalt doped proangiogenic-osteogenic hydroxyapatite. Hydroxyapatite samples, doped with varying concentrations of bivalent cobalt (Co(2+)) were prepared by the ammoniacal precipitation method and the extent of doping was measured by ICP-OES. The
Kalimuthu Pandi et al.
Carbohydrate polymers, 134, 732-739 (2015-10-03)
Adsorption is a significant reaction occurs between adsorbent/water interface for controlling the pollutants in the aqueous environment. In this regard, an eco-magnetic biosorbent was prepared by uniform deposition of magnetic Fe3O4 particles on the surface of nano-hydroxyapatite (n-HAp)/chitosan (CS) nanocomposite
Cong Mao et al.
Biomedical materials (Bristol, England), 10(2), 025005-025005 (2015-03-26)
The ability of biomaterials to induce rapid vascular formation is critical in tissue regeneration. Combining recombinant angiogenic growth factors with bioengineered constructs have proven to be difficult due to several issues, including the instability of recombinant proteins, the need for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service