Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-3170

Sigma-Aldrich

Brij® L23

Synonym(s):

Brij 35, C12E23, Polyoxyethylene (23) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

availability

available only in Japan

technique(s)

microbiological culture: suitable
protein quantification: suitable

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

Inchi Key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Suitability

amino acid analysis

Legal Information

Brij is a registered trademark of Croda International PLC

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramon R J P van Eekeren et al.
Journal of vascular surgery, 57(2), 445-450 (2012-11-13)
Thermal ablative techniques of varicose veins carry a risk of heat-related complications, including postoperative pain. Mechanochemical endovenous ablation (MOCA) might avoid these complications and reduce postoperative pain because of the absence of thermal energy. This study evaluated postoperative pain and
Adriana Smarandache
Photomedicine and laser surgery, 30(5), 262-267 (2012-03-16)
The goal of this study was to investigate the laser light scattering involved in the laser irradiation process of polidocanol foam samples. Clinical experimental results proved that exposure of tissues impregnated with foaming polidocanol to laser radiation emitted at 1064
Albert-Adrien Ramelet
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(5), 748-751 (2012-01-25)
Some leg telangiectasias may be refractory to treatment, including sclerotherapy and lasers. To describe the innovative Sclerotherapy in Tumescent Anesthesia of Reticular veins and Telangiectasias (START) approach to achieving good results in such patients, which also proves effective in treating
G C Valenzuela et al.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery, 46(5), 593-599 (2013-09-28)
Sclerosant foams are aqueous and break down under the influence of gravity, pressure, and temperature. The aim of this study was to investigate the effects of temperature on foam stability. Sodium tetradecyl sulphate (STS) and polidocanol (POL) liquid and foam
Sabrina G Fabi et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 38(7 Pt 2), 1112-1127 (2012-01-25)
The aged hand is characterized by cutaneous and dermal atrophy, with deep intermetacarpal spaces, prominent bones and tendons, and bulging reticular veins. Epidermal changes include solar lentigines, seborrheic keratoses, actinic keratoses, skin laxity, rhytides, tactile roughness, and telangiectasia. A Medline

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service