Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-1720

Sigma-Aldrich

Benzyl benzoate

SAJ special grade, ≥99.0%

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
Beilstein/REAXYS Number:
2049280
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

vapor pressure

1 mmHg ( 125 °C)

assay

≥99.0%

autoignition temp.

896 °F

availability

available only in Japan

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

298.4 °F

flash_point_c

148 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing-Liang Li et al.
The journal of physical chemistry. B, 113(15), 5011-5015 (2009-03-25)
The formation of most organogels requires the compatibility of both the gelator and solvent. It is very desirable if the rheological properties of a gel can be manipulated to achieve the desired performance. In this paper, a novel organogel was
Glen H MacDonald et al.
Hearing research, 243(1-2), 1-10 (2008-06-25)
The complex anatomy of the mammalian cochlea is most readily understood by representation in three-dimensions. However, the cochlea is often sectioned to minimize the effects of its anatomic complexity and optical properties on image acquisition by light microscopy. We have
Ben Spitzer-Rimon et al.
The Plant cell, 22(6), 1961-1976 (2010-06-15)
Floral scent, which is determined by a complex mixture of low molecular weight volatile molecules, plays a major role in the plant's life cycle. Phenylpropanoid volatiles are the main determinants of floral scent in petunia (Petunia hybrida). A screen using
Sandrine Joly et al.
The European journal of neuroscience, 40(7), 3021-3031 (2014-07-22)
The lack of axonal regeneration in the adult central nervous system is in part attributable to the presence of inhibitory molecules present in the environment of injured axons such as the myelin-associated proteins Nogo-A and MAG and the repulsive guidance
Catherine W McCollum et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 152-163 (2014-04-29)
Exposure to arsenic in its inorganic form, arsenite, causes adverse effects to many different organs and tissues. Here, we have investigated arsenite-induced adverse effects on vascular tissues in the model organism zebrafish, Danio rerio. Zebrafish embryos were exposed to arsenite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service