Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03-0675

Sigma-Aldrich

Beeswax

yellow, CP

Synonym(s):

Cera alba

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12181502
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

color

yellow

mp

61-65 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter A Lewis et al.
Journal of burn care & research : official publication of the American Burn Association, 33(4), e195-e200 (2012-06-06)
Postburn itch is reported to affect up to 87% of the burn population. Although treatments for postburn itch are multimodal, they remain consistently ineffective. However, recent anecdotal evidence from several outpatients at a tertiary referral hospital suggests that a cream
M Cristina Gamberini et al.
Analytical and bioanalytical chemistry, 401(6), 1839-1846 (2011-07-05)
A study of the composition of the remains of ancient ointments from museums was undertaken to enable understanding of the preparation techniques. Comparison of ancient recipes from different historical periods and spectroscopic characteristics of inorganic and/or organic remains recovered in
Christian W W Pirk et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 181(3), 353-359 (2010-11-17)
Colonies of Apis florea, which only abscond a short distance, usually return to salvage old nest wax; but, those colonies, and all other honeybee species which go considerably further, do not. Wax salvage would clearly be counter-productive unless the energy
Chiara Riedo et al.
Analytical and bioanalytical chemistry, 401(6), 1761-1769 (2011-06-04)
Analytical pyrolysis with thermally assisted hydrolysis and methylation was employed to investigate ancient ointments collected from Spanish vessels coming from the sixteenth century pharmacies. The ointments were reproduced on the basis of historical recipes and characterization was made in comparison
R J Stacey
Analytical and bioanalytical chemistry, 401(6), 1749-1759 (2011-06-18)
Residues from medicine containers in the collections of the British Museum have been investigated as part of a wider programme of scientific work on Roman surgical instruments. The cylindrical bronze containers are often described as instrument cases, but some contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service