Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-6190

Sigma-Aldrich

Antimony(III) oxide

CP

Synonym(s):

Diantimony trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2O3
CAS Number:
Molecular Weight:
291.52
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

form

solid

reaction suitability

reagent type: catalyst
core: antimony

availability

available only in Japan

bp

1550 °C (lit.)

mp

655 °C (lit.)

SMILES string

O=[Sb]O[Sb]=O

InChI

1S/3O.2Sb

InChI key

ADCOVFLJGNWWNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Kirkland et al.
Mutation research, 627(2), 119-128 (2006-12-19)
Antimony trioxide (Sb2O3, CAS 1309-64-4) is widely used as a flame retardant synergist in a number of household products, as a fining agent in glass manufacture, and as a catalyst in the manufacture of various types of polyester plastics. It
Koen Oorts et al.
Environmental science & technology, 42(12), 4378-4383 (2008-07-09)
Antimony trioxide (Sb2O3) is a widely used chemical that can be emitted to soil. The fate and toxicity of this poorly soluble compound in soil is insufficiently known. A silt-loam soil (pH 7.0, background 0.005 mmol Sb kg(-1)) was amended
Nona Merry M Mitan et al.
Chemosphere, 72(7), 1073-1079 (2008-05-24)
The controlled pyrolysis of polyethylene/polypropylene/polystyrene mixed with brominated high-impact polystyrene containing decabromodiphenyl ether as a brominated flame-retardant with antimony trioxide as a synergist was performed. The effect of decabromodiphenyl ether and antimony trioxide on the formation of its congeners and
Lisa Bregoli et al.
Toxicology, 262(2), 121-129 (2009-06-02)
Nanoparticles (NPs) are materials with one dimension in the range of 1-100 nm. The toxicity of NPs remains widely unknown and still poses concerns, due to the peculiar characteristics of materials in the nano-size range. We analyze the toxicity of
Susan Lösler et al.
Annals of hematology, 88(11), 1047-1058 (2009-03-21)
During the last years remission rates of more than 72% for arsenic(III)-oxide (As(2)O(3)) treatment in relapsed or refractory acute promyelocytic leukemia have been published. As(2)O(3) is under clinical investigation for therapy of leukemia and solid tumors. Due to the chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service