Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-2660

Sigma-Aldrich

Aluminum chloride hexahydrate

JIS special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3 · 6H2O
CAS Number:
Molecular Weight:
241.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

vapor pressure

1 mmHg ( 100 °C)

assay

≥98.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: aluminum

availability

available only in Japan

pH

2.5-3.5 (20 °C)

SMILES string

Cl[Al](Cl)Cl.O.O.O.O.O.O

InChI

1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3

InChI key

JGDITNMASUZKPW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

target_organs

Lungs

supp_hazards

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aisha Nasir Hashmi et al.
The International journal of neuroscience, 125(4), 277-287 (2014-05-16)
Aluminium metal (Al) has been implicated in the etiology of many neurodegenerative diseases, most commonly the Alzheimer's disease (AD). Al causes biochemical defects by affecting the neurotransmitters level, causes conformational changes in amyloid β protein and increases amyloid accumulation in
Ji-Won Jung et al.
ACS nano, 9(7), 6717-6727 (2015-06-02)
Amorphous silicon (a-Si) has been intensively explored as one of the most attractive candidates for high-capacity and long-cycle-life anode in Li-ion batteries (LIBs) primarily because of its reduced volume expansion characteristic (∼280%) compared to crystalline Si anodes (∼400%) after full
Dragana M Vucić et al.
Acta poloniae pharmaceutica, 70(6), 1021-1026 (2014-01-05)
Antibacterial and antioxidant activity, total phenolic and flavonoid concentrations of aqueous, ethanol and ethyl acetate extracts from the leaves and flowers of Erica herbacea L. were studied. In vitro antibacterial activity of the extracts was determined by macrodilution method. Minimum
Syed Husain Mustafa Rizvi et al.
PloS one, 9(5), e98409-e98409 (2014-06-01)
Aluminium (Al) is the third most abundant element in the earth's crust and its compounds are used in the form of house hold utensils, medicines and in antiperspirant etc. Increasing number of evidences suggest the involvement of Al+3 ions in
May S M Wee et al.
Carbohydrate polymers, 130, 57-68 (2015-06-17)
The shear-thickening rheological behaviour (between 5 and 20s(-1)) of a 5% (w/w) viscoelastic gum extracted from the fronds of the native New Zealand black tree fern or mamaku in Māori was further explored by manipulating the salt content. The freeze-dried

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service