Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAEIII-RO

Roche

Mae III

from Methanococcus aeolicus PL-15/H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

bacterial (Methanococcus aeolicus PL-15/H)

form

solution

packaging

pkg of 250 U (10822248001 [1 - 5 U/μl])
pkg of 50 U (10822230001 [1 - 5 U/μl])

manufacturer/tradename

Roche

parameter

55 °C optimum reaction temp.

storage temp.

−20°C

General description

Mae III is a restriction enzyme which recognizes the sequence ↓GTNAC and generates fragments with 5′-cohesive termini.[1]

Specificity

Recognition sites: GTNAC
GTNAC
Restriction site: ↓GTNAC
↓GTNAC
Heat inactivation: There is no information available whether or not Mae III can be heat inactivated.

Application

Mae III has been used as a restriction enzyme for PCR products.[2]

Quality

Absence of nonspecific endonuclease activities
1μg λDNA is incubated for 16hours in 50μl incubation buffer with an excess of Mae III. The number of enzyme units which do not change the enzyme-specific pattern is stated in the certificate of analysis.

Absence of exonuclease activity

Approximately 5μg [3H] labeled calf thymus DNA are incubated with 3μl Mae III for 4hours at +37°C in a total volume of 100μl 50mM Tris-HCl, 10mM MgCl2, 1mM Dithioerythritol, pH approximately 7.5. Under these conditions, no release of radioactivity is detectable, as stated in the certificate of analysis.

DNA Profile

Number of cleavage sites on different DNAs
  • λ: 156
  • φX174: 17
  • Ad2: 118
  • M13mp7: 25
  • pBR322: 17
  • pBR328: 18
  • pUC18: 11
  • SV40: 14

Unit Definition

One unit is the enzyme activity that completely cleaves 1 μg λDNA in one hour at +55 °C in a total volume of 25 μl (1x) special Mae I incubation buffer.

Analysis Note

Compatible ends
Mae III ends are compatible with ends generated by Bst E II.

Isoschizomers

The enzyme has no known isoschizomers.

Methylation sensitivity
There is no evidence that the enzyme is inhibited by methylation.

SuRE/Cut Buffer System

The buffer in bold is recommended for optimal activity
  • A: 0-10%
  • B: 10-25%
  • H: 10-25%
  • L: 0-10%
  • M: 0-10%
Note: Instead of the SuRE/Cut buffer system, please use the special 2x incubation buffer supplied with the enzyme.

Incubation temperature
+55°C

Unit definition

One unit is the enzyme activity that completely cleaves 1μg λDNA in 1 hour at +55°C in a total volume of 25μl special incubation buffer.

Heat inactivation

No information available.

Ligation and recutting assay
Mae III fragments obtained by complete digestion of 1μg λDNA are ligated with 1U T4 DNA Ligase in a volume of 10μl by incubation for 16 hours at +4°C in 66mM Tris-HCl, 5mM MgCl2, 5mM Dithiothreitol, 1mM ATP, pH 7.5 (at +8°C) resulting in >95% recovery of 1μg λDNA fragments.
Subsequent re-cutting with Mae III yields >95% of the typical pattern of λDNA × Mae III fragments.
Activity in PCR buffer: 0%

Relative activity in PCR mix (Taq DNA Polymerase buffer) is 0%. The PCR mix contained λDNA, primers, 10 mM Tris-HCl (pH 8.3, 20 °C), 50 mM KCl, 1.5 mM MgCl2, 200 μM dNTPs, 2.5 U Taq DNA polymerase. The mix was subjected to 25 amplification cycles.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Enzyme Solution

  • Special incubation buffer for Mae III

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Jacobson et al.
American journal of human genetics, 47(1), 127-136 (1990-07-01)
Senile systemic amyloidosis (SSA) is a late-onset disease characterized by deposition of amyloid fibrils containing transthyretin (TTR). Amino acid sequencing of protein isolated from the amyloid fibrils of a patient with SSA identified TTR containing a position - 122 isoleucine-for-valine
Siobhan Eustace Ryan et al.
Eye and vision (London, England), 2, 18-18 (2015-11-26)
Leber's Hereditary Optic Neuropathy (LHON; MIM 535000) is one of the most commonly inherited optic neuropathies and it results in significant visual morbidity among young adults with a peak age of onset between the ages of 15-30. The worldwide incidence

Articles

The term “Restriction enzyme” originated from the studies of Enterobacteria phage λ (lambda phage) in the laboratories of Werner Arber and Matthew Meselson.

Related Content

Restriction endonucleases in prokaryotes function primarily to protect against foreign genetic material, notably bacteriophage DNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service