Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HIST-RO

Roche

Histone H1

from calf thymus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

lyophilized

packaging

pkg of 1 mg (10223549001)
pkg of 5 mg (11004875001)

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

Histone H1 plays an essential role in maintaining and establishing higher order chromatin structures. The linker histone H1 basically binds to the DNA which is entering and exiting the nucleosomal core particle. All H1 variants in higher eukaryotes comprises of a general structure, consisting of a central conserved globular domain and less conserved N-terminal and C-terminal tails. It has been suggested that each H1 variant has a specific function owing to its tail region which is specifically conserved among species, but differ in variants.[1][2]

Application

Lysine-rich preparation used as a substrate for protein kinases.

Biochem/physiol Actions

Histone H1 was used as a substrate in kinase assay.[1][2]
Studies have suggested that histoneH1 might act as a general repressor of transcription.[1][2]

Quality

Typical analysis: The preparation is electrophoretically homogeneous.
Typical analysis: The preparation is electrophoretically homogeneous.

Sequence

Histone H1 consists of 212 amino acids.
Histone H1 has a very high lysine content (approx. 25%) which contributes to its high basic charge.

Preparation Note

Working concentration: 1 to 5 mg/ml
Working solution: Solvent is recommended in double-distilled water.
Storage conditions (working solution): Aqueous solution of Histone H1 is stable at -15 to -25 °C for at least three months.
Avoid repeated freezing and thawing!

Other Notes

For life science research only. Not for use in diagnostic procedures.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annalisa Izzo et al.
Biological chemistry, 389(4), 333-343 (2008-01-23)
The linker histone H1 binds to the DNA entering and exiting the nucleosomal core particle and has an important role in establishing and maintaining higher order chromatin structures. H1 forms a complex family of related proteins with distinct species, tissue
Boris Kysela et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(6), 1877-1882 (2005-01-27)
DNA nonhomologous end-joining in vivo requires the DNA-dependent protein kinase (DNA-PK) and DNA ligase IV/XRCC4 (LX) complexes. Here, we have examined the impact of histone octamers and linker histone H1 on DNA end-joining in vitro. Packing of the DNA substrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service