Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G50DNA-RO

Roche

Quick Spin Columns for radiolabeled DNA purification

Sephadex G-50, fine

Synonym(s):

DNA purification, spin column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

packaging

pkg of 20 columns (11273965001)
pkg of 50 columns (11273973001)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Ready-to-use disposable columns designed to quickly and efficiently remove unincorporated precursors from labeled DNA. Quick Spin columns are designed for use in low-speed, swinging-bucket centrifuges.Quick Spin Columns are produced with material shown to have high recovery of radiolabeled DNA (≥90%).

  • All the tedious, time-consuming steps involved in preparing columns have already been performed. The columns are pre-packed, pre-swollen, and quality tested to ensure maximal retention of unincorporated nucleotides (≥99%).

Contents
Pre-packed, pre-swollen, pre-spun columns.
Suspension of Sephadex G-50 in STE buffer (10 mM Tris-HCl, pH 7.5, 1 mM EDTA, 100 mM NaCl).

Application

Remove unincorporated precursors from DNA labeled by nick translation, end labeling, polymerization reactions, and other labeling techniques.
Use the highly purified radiolabeled DNA in Southern blotting and other downstream applications.

Features and Benefits

  • Save time and effort with ready-to-use columns.
  • Increase yields of nucleic acid in less time and with fewer steps.
  • Ensure optimal performance and reproducibility with quality tested columns:
a)High recovery of radiolabeled DNA
b)Maximum retention of unincorporated nucleotides
c)Absence of DNases

Quality

DNases are not detected, according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service