Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G25DNA-RO

Roche

Quick Spin Columns for radiolabeled DNA purification

Sephadex G-25, fine

Synonym(s):

DNA purification, spin column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

packaging

pkg of 20 columns (11273922001)
pkg of 50 columns (11273949001)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Contents
Pre-packed, pre-swollen, pre-spun columns.
Suspension of Sephadex G-25 in STE buffer (10 mM Tris-HCl, pH 7.5, 1 mM EDTA, 100 mM NaCl).
Quick Spin columns are produced with material that is shown to have a high recovery of radiolabeled DNA ( ≥80%).
All tedious, time-consuming steps involved in preparing columns have already been performed. The columns are pre-packed and quality tested to ensure
  • maximal retention of unincorporated nucleotides (≥95%), and
  • absence of DNase contamination according to the current Quality Control procedures.
Quick Spin columns contain gel filtration matrices which allow large molecules (e.g., DNA or RNA) to pass through quickly while retaining small molecules (e.g., nucleotides). The Quick Spin format improves the molecular sieving concept by using centrifugation to separate DNA or RNA rapidly and cleanly from small contaminants.

Specificity

Exclusion limit: 10-12 bp

application

Ready-to-use disposable column units designed to quickly and efficiently remove unincorporated precursors from DNA labeled by nick translation, end-labeling, polymerization reactions, and other labeling techniques.
Quick Spin columns are designed for use in low-speed, swinging-bucket centrifuges.

Analysis Note

Sample size: up to 50 μl
Time required: 10 minuntes

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service