Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11988506001

Roche

Anti-HA-Fluorescein, High Affinity

from rat IgG1

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

rat

conjugate

fluorescein conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F10, monoclonal

form

lyophilized

packaging

pkg of 25 μg

manufacturer/tradename

Roche

isotype

IgG1

epitope sequence

YPYDVPDYA

storage temp.

−20°C

General description

Anti-HA-Fluorescein, High Affinity is a monoclonal antibody for the highly sensitive detection of HA-tagged recombinant proteins, Fab fragments, conjugated to fluorescein.
Anti-HA-Fluorescein, High Affinity recognizes the HA peptide sequence (YPYDVPDYA), derived from the human hemagglutinin protein. The antibody recognizes its antigenic determinant even when the HA peptide epitope is introduced into unrelated recombinant proteins by a technique known as "epitope tagging".

Contents
Lyophilizate, stabilized

Specificity

Anti-HA-Fluorescein, High Affinity (3F10) recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein. This epitope is also recognized in fusion proteins regardless of its position (N-terminal, C-terminal or internal).

Immunogen

Amino acids 98 to 106 from the human influenza virus hemagglutinin protein

Application

Anti-HA-Fluorescein, High Affinity has been used in immunofluorescence labeling of chinese hamster ovary live cells[1] and in immunocytochemistry of neuron cell culture.[2]

Quality

The Anti-HA-Fluorescein, High Affinity antibody is function tested by flow cytometry analysis of a HA-tagged fusion protein

Preparation Note

Stabilizers: proteinous stabilizers
Working concentration: Flow cytometry: 1 to 5 μg/ml
Immunofluorescence: 1 to 5 μg/ml
Working concentration of conjugate depends on application and substrate. Concentrations should be taken as a guideline.
Storage conditions (working solution): Reconstituted antibody is stable at 2 to 8 °C for two months. Alternatively store aliquots at -15 to -25 °C for up to six months.
Note: Avoid thawing and repeated freezing.

Reconstitution

Add 0.5 ml double-distilled water to a final concentration of 50 μg/ml. Rehydrate for 10 minutes prior to use.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Toshiya Ando et al.
Current biology : CB, 29(9), 1512-1520 (2019-04-23)
Nanometer-level patterned surface structures form the basis of biological functions, including superhydrophobicity, structural coloration, and light absorption [1-3]. In insects, the cuticle overlying the olfactory sensilla has multiple small (50- to 200-nm diameter) pores [4-8], which are supposed to function
Alfred J Zullo et al.
Journal of immunology (Baltimore, Md. : 1950), 178(1), 58-66 (2006-12-22)
NKT cells are glycolipid-reactive lymphocytes that express markers and perform functions common to both T lymphocytes and NK cells. Although the genetic events controlling conventional T cell development are well defined, the transcription factors and genetic programs regulating NKT cell
Enhanced dimerization drives ligand-independent activity of mutant epidermal growth factor receptor in lung cancer.
Valley CC, et al.
Molecular Biology of the Cell, 26(22), 4087-4099 (2015)
A combined transgenic proteomic analysis and regulated trafficking of neuroligin-2.
Kang Y, et al.
The Journal of Biological Chemistry, 289(42), 29350-29364 (2014)
Christopher C Valley et al.
Molecular biology of the cell, 26(22), 4087-4099 (2015-09-05)
Mutations within the epidermal growth factor receptor (EGFR/erbB1/Her1) are often associated with tumorigenesis. In particular, a number of EGFR mutants that demonstrate ligand-independent signaling are common in non-small cell lung cancer (NSCLC), including kinase domain mutations L858R (also called L834R)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service