Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11855646001

Roche

DNA Molecular Weight Marker XVII (500 bp ladder)

pkg of 50 μg (in 200 μl), solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

packaging

pkg of 50 μg (in 200 μl)

manufacturer/tradename

Roche

storage temp.

−20°C

General description

Fragment mixture prepared by cleavage of a specially constructed plasmid with restriction endonucleases.
Size Range: 0.5 to 5.0 kbp.

Application

DNA Molecular Weight Marker XVII (500 bp ladder) allows accurate sizing of DNA fragments generated by PCR,[1] RT-PCR or restriction digest, following separation on agarose gels. The DNA Molecular Weight Marker XVII can be used in conjunction with DNA Molecular Weight Marker XIII (50 bp ladder), DNA Molecular Weight Marker XIV (100 bp ladder), and DNA Molecular Weight Marker XVI (250 bp ladder) for precise size determination.

Sequence

The mixture contains 10 blunt-ended, double-stranded DNA fragments with the following base pair lengths (1 base pair = 660 daltons): 500, 1000, 1500, 2000, 2500, 3000, 3500, 4000, 4500, and 5000. The 2000- and 4000-bp banding pattern are three times brighter. Electrophoretic separation of this molecular weight marker results in a regular pattern.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels. The fragments exhibit uniform spacing for easier reading, while bands 2000 and 4000 are 3 times brighter for easy orientation.

Unit Definition

50 μg = 1A260 unit

Physical form

Ready-to-use solution, 250μg/ml, in TE buffer (10mM Tris-HCl, 1mM EDTA, pH 8.0).

Preparation Note

Storage conditions (working solution): 2 to 8 °C
Once thawed, store at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlene R Jackson et al.
Applied and environmental microbiology, 70(7), 4205-4210 (2004-07-09)
The effect of tylosin on erythromycin-resistant enterococci was examined on three farms; farm A used tylosin for growth promotion, farm B used tylosin for treatment of disease, and farm C did not use tylosin for either growth promotion or disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service