Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11242008001

Roche

L-Carnitine

sufficient for ~3 x 10 tests

Synonym(s):

L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for ~3 x 10 determinations (test-combination)
sufficient for ~3 x 10 tests

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Enzymatic UV-Test with a linearity in the range of 5.6 - 112 μM L-carnitine in the test solution.

Application

Determination of L-carnitine in seminal plasma, serum, or urine in life science research applications.

Biochem/physiol Actions

L-carnitine is an abundant amino acid present in red meat. Its trimethylamine structure has similarity to that of choline. The major source of this amino acid is by dietary ingestion. In mammals, it is also endogenously produced from lysine. It participates in the transport of fatty acids into the mitochondrial compartment. It produces trimethylamine-N-oxide (TMAO) and promotes atherosclerosis.

Packaging

1 kit containing 6 components

Principle

L-Carnitine is acetylated to acetyl carnitine by acetyl coenzyme A (acetyl CoA) in the presence of the enzyme carnitine acetyl transferase (CAT). The resulting coenzyme A (CoA) is acetylated back to acetyl CoA in the presence of adenosine-5′-triphosphate (ATP) and acetate, catalyzed by the enzyme acetyl CoA synthetase (ACS). This results in the formation of adenosine-5′-monophosphate (AMP) and inorganic pyrophosphate (PPi). In the presence of ATP, supported by myokinase (MK), AMP forms twice the amount of adenosine-5′-diphosphate (ADP). This is converted in the following reaction with phosphoenol pyruvate (PEP) in the presence of pyruvate kinase (PK). The pyruvate formed is reduced to L-lactate by reduced nicotinamide adenine dinucleotide (NADH) in the presence of lactate dehydrogenase (LDH). The amount of NADH consumed during the reaction is equivalent to half the amount of L-carnitine. NADH is measured spectroscopically at 340 nm.

Preparation Note

Storage conditions (working solution): Note:
  • After reconstitution, the solution 1 is stable for 4 days at 2 to 8 °C.
  • Bring solution 1 to 15 to 25 °C prior to use.
  • After reconstitution, the suspension 2 is stable for 3 months at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Coenzyme/Buffer Mixture

  • Enzyme Component A: Acetyl-CoA synthetase

  • Enzyme blend B: Enzyme suspension with myokinase, lactate dehydrogenase, and pyruvate kinase

  • Carnitine acetyltransferase

  • L-Carnitine Standard Solution

  • Detergent Solution

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Correlation between Plasma L-carnitine and Growth Performance in Suckling Pigs in Respect of mRNA Expression of a Carnitine Transporter, OCTN2, in Lactating Porcine Mammary Tissues.
Thongsong B and Thongsong SK
Thai Journal of Veterinary Medicine, 38, 27-36 (2008)
Ke Li et al.
Journal of chromatographic science, 48(5), 371-374 (2010-06-03)
A simple and reliable high-performance liquid chromatographic (HPLC) method for simultaneous determination of free and total carnitine in human serum has been developed, and its clinical significance has been investigated. After proteins in serum were precipitated, carnitine in serum was
Intestinal microbiota metabolism of L-carnitine, a nutrient in red meat, promotes atherosclerosis.
Koeth RA, et al.
Nature Medicine, 19, 576-585 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service