Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10821705001

Roche

DNA Molecular Weight Marker V

pkg of 50 μg (in 200 μl), solution

Synonym(s):

DNA Marker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335

form

solution

packaging

pkg of 50 μg (in 200 μl)

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

General description

Fragment mixture prepared by cleavage of pBR322 DNA with restriction endonuclease Hae III.

Size Range: 8 to 587 bp

Application

DNA Molecular Weight Marker V is used for the size determination of DNA in agarose gels. It simplifies accurate molecular weight determination of double-stranded DNA fragments generated by:

<UL><LI>Restriction digests</LI>

<LI>PCR</LI> [1]

<LI>RT-PCR</LI></UL>

Components

Ready-to-use solution, 250μg/ml, in TE buffer (10mM Tris-HCl, 1mM EDTA, pH 8.0)

Sequence

The mixture contains 22 DNA fragments with the following base pair lengths (1 base pair = 660 daltons): 8, 11, 18, 21, 51, 57, 64, 80, 89, 104, 123, 124, 184, 192, 213, 234, 267, 434, 458, 504, 540, and 587 as determined by computer analysis of the pBR322 sequence.
Note: Fragment lengths are derived from computer analysis of the DNA sequence. Depending on the size range of the marker, the smallest fragments will only be visible on overloaded gels.

Unit Definition

50 μg = 1A260 unit

Preparation Note

Activator: Ammonia, pyridine, imidazole, all at pH >7; detergents

Working concentration: A suggested working ratio, for immediate use in coupling procedures, is 2 mg POD to 4 mg lgG (molar ratio = 2:1, POD:IgG).

Storage conditions (working solution): 2 to 8 °C
Once thawed we recommend further storage at 2 to 8 °C.

Storage and Stability

Store at -15–-25 °C. (until use)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Romero et al.
Nature protocols, 3(11), 1729-1735 (2008-10-18)
The lamin B2 locus is the only mammalian origin whose replication initiation points (RIPs) have been mapped. Although this paper was published 8 years ago, no further mammalian RIP-mapping studies have been reported, largely due to technical difficulties of ligation-mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service