04743814001
Roche
Expand™ 20 kbPLUS PCR System, dNTPack
sufficient for ≤40 reactions, pkg of 200 U, suitable for PCR, dNTPs included
About This Item
Recommended Products
usage
sufficient for ≤40 reactions
feature
Long Range PCR
dNTPs included
hotstart: no
packaging
pkg of 200 U
manufacturer/tradename
Roche
storage condition
avoid repeated freeze/thaw cycles
parameter
68 °C optimum reaction temp.
technique(s)
PCR: suitable
input
purified DNA
shipped in
dry ice
storage temp.
−20°C (−15°C to −25°C)
General description
T/A cloning can also be performed.
Use 3.75 U for a standard 50μL PCR.
Each dNTPack contains 10mM additive-free sodium salt nucleotides as a ready-to-use mix.
To obtain long PCR products, first evaluate length and purity of the DNA template. Handling all genomic DNA templates with care. Genomic DNA template should be larger than 50kb, as determined with agarose gel electrophoresis.
Application
- PCR
- RT-PCR
- Large-fragment amplification
Features and Benefits
- Go the limit: The buffer and enzyme-blend amplify products over 20kb DNA.
- Easy PCR monitoring: Human genomic DNA and human control β-globin primers amplify a 23kb fragment.
- Test template quality: Control reagents also test template quality and primer pair performance.
- Cost-effective: Use the convenient premixed solution of PCR grade dNTPs.
Packaging
Quality
Unit Definition
Storage and Stability
Other Notes
Legal Information
Kit Components Only
- Expand 20 kbPLUS Enzyme Mix
- Expand 20 kbPLUS Reaction Buffer, with 27.5 mM MgCl2 10x concentrated
- MgCl2 Stock Solution 25 mM
- Human Genomic DNA
- Human β-globin Control Primer forward, (HβG forward)
- Human β-globin Control Primer reverse, (HβG reverse)
- PCR Nucleotide Mix
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 3
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_c
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The purpose of Hot Start PCR is to inhibit the PCR reaction in order to reduce nonspecific amplification, prevent the formation of primer dimers, and increase product yields.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service