Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03117928001

Roche

Quick Spin High Capacity Columns

pkg of 20 columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Product 03117928001 is not currently sold in your country. Contact Technical Service

description

Sephadex G-50, fine

Quality Level

packaging

pkg of 20 columns

manufacturer/tradename

Roche

General description

Quick Spin High Capacity Columns are produced with material shown to have recovery of DNA (≥90%). All the tedious, time-consuming steps involved in preparing columns have already been performed. The columns are pre-swollen, pre-packed, pre-spun, and quality tested to ensure
  • maximal retention of unincorporated nucleotides (≥98%), and
  • absence of DNase contaminaton according to the current Quality Control procedures.

Application

Quick Spin High Capacity G-50 Sephadex columns has been used to desalt viral samples.[1] It has been used during the purification process in order to remove humic substances from RNA samples.[2]

Features and Benefits

Exclusion limit for ds DNA: >72 base pairs
Recovery: =90% of DNA
Retention: =98% of unincorporated nucleotides
Sample size: 100 to 500 μl

Physical form

Pre-packed, pre-swollen, columns (3.0 ml bed volume). Suspension of Sephadex G-50 in TE buffer (10 mM Tris, pH 8.0; 1 mM EDTA).

Storage and Stability

Store at 2 to 8 °C. (Do not freeze. To avoid contamination of the columns and collecting tubes, leave them in the storage bag until just before use.)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Wang et al.
Bioscience, biotechnology, and biochemistry, 72(3), 694-701 (2008-03-08)
To understand the response of soil bacteria to the surrounding environment, it is necessary to examine the gene expression profiles of the bacteria in the soil. For this purpose, we developed a new method of extracting RNA from soil reproducibly.
Resham Bhattacharya et al.
The Journal of biological chemistry, 280(32), 28848-28851 (2005-06-28)
The Kruppel-like factor KLF2 was recently identified as a novel regulator of endothelial pro-inflammatory and pro-thrombotic function. Here it is shown that overexpression of KLF2 potently inhibits vascular permeability factor/vascular endothelial growth factor (VEGF-A)-mediated angiogenesis and tissue edema in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service