Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

YY2004K00-M

ESP valve Plugs kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
Pricing and availability is not currently available.

shipped in

ambient

General description

Microfil devices offer a convenient monitoring method to detect contamination related problems easily and reliably.

Components

3 fluoroelastomer o-rings

5 syringe needles

10 plastic caps

3 plug seals

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service