Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y04E-01-5PK

CELLASIC® ONIX plate diploid yeast cells (4 chamber, 5-7 micron)

Synonym(s):

4-Chamber Yeast Plate, Diploid Yeast Plate, ONIX Yeast Plate

Sign Into View Organizational & Contract Pricing

Select a Size

Y04E-01-5PK
$1,220.00

$1,220.00


Check Cart for Availability

Request more information

Select a Size

Change View
Y04E-01-5PK
$1,220.00

About This Item

UNSPSC Code:
41161500

$1,220.00


Check Cart for Availability

Request more information

material

PMMA
glass
polycarbonate
polydimethylsiloxane (PDMS)

packaging

pack of 5 ea

L × W × H

127.76 mm × 85.48 mm × 14.35 mm

General description

Cell size range: 5.0-7.0 μm
Device configuration: 4 chamber
Tiered ceiling (5.0, 6.0, 7.0 μm) limits cell movement in the Z-plane to retain cells within the field of view.

Application

The Y04E plates for Diploid Yeast Cells utilize a microfabricated silicone ceiling with a height similar to yeast cells to restrict their growth in a single focal plane & maintaining x,y position over time.

Other Notes

Direct replacement to Y04D-02-5PK for diploid yeast cell culture.

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service