TX1282P-1 is packaged in poly coated glass bottles
Preparation Note
Product filtered through a 0.2 μm filter
Analysis Note
Assay (trifluoroacetic acid)0.045-0.055% (v/v)Color (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidGradient at 210 nm0.01AU maxGradient at 220 nm0.001AU maxGradient at 280 nm0.001AU maxUV Abs. at 205 nm1.00AU maxUV Abs. at 210 nm0.70AU maxUV Abs. at 230 nm0.30AU maxUV Abs. at 250 nm0.05AU maxUV Abs. at 280 nm0.010AU maxUV Abs. at 300 nm0.010AU maxUV Abs. at 350 nm0.010AU max
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.