Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TTHA0F210

Millipore

Sterisure filter device, mixed cellulose esters membrane 0.45 µm

The Sterisure filter device with mixed cellulose esters membrane is a sterilized, disposable filtration device which is used for testing pharmaceuticals and biopharmaceuticals in an Isolator.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

agency

USP 71

Quality Level

manufacturer/tradename

Sterisure

input

sample type pharmaceutical(s)

application(s)

sterility testing

shipped in

ambient

General description

Use Sterisure devices to test pharmaceuticals and biopharmaceuticals in an isolator. The double packaging reduces particulate transfer into the isolator or into the designated sterility testing area. The outer Sterisure package holds 10 individual packages, each containing a pair of sterilized devices. Versatile design lets you test low protein binding agents, medical devices (by direct inoculation), and diagnostic test products for the presence/absence of microorganisms. May also be used for microbial limits testing with the Milliflex PLUS pump.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service