Key Documents
TRCSAU0PCERIL
Operator Certification for Automatic Filter Integrity Testing (at Customer Site)
Select a Size
TRCSAU0PCER
$13,439.00
$13,439.00
Check Cart for Availability
Select a Size
Change View
TRCSAU0PCER
$13,439.00
About This Item
UNSPSC Code:
86132100
$13,439.00
Check Cart for Availability
General description
Using a combination of theory and hands-on instruction, you will learn the key procedures of filter integrity testing, how to read printouts from automatic filter-integrity testers and troubleshoot filter-integrity test processes. This two-day course covers all current filter integrity tests, including bubble point, diffusive flow and water-flow integrity testing that apply to all liquid and gas membrane filtration applications.
Visit the Document Library
Related Content
Pharmaceutical and biopharmaceutical manufacturers can enhance their skill sets with practical, hands-on training courses in-person or streamed live from our M Lab™ Collaboration Centers.