Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX1015

Supelco

Strontium Chloride, Hexahydrate

Meets ACS Specifications GR ACS

Synonym(s):

Strontium Chloride, Hexahydrate, Strontium Chloride Hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Sr · 6H2O
CAS Number:
Molecular Weight:
266.62
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-971-6

Quality Level

assay

99.0-103.0% (silver nitrate titration)

form

solid

impurities

≤0.005% Insoluble matter

pH

5-7 (25 °C, 50 g/L in H2O)

mp

61 °C

solubility

2058 g/L

anion traces

sulfate (SO42-): ≤0.001%

cation traces

Ba: ≤0.05%
Ca: ≤0.05%
Fe: ≤5 ppm
Mg: ≤2 ppm
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2

InChI key

AMGRXJSJSONEEG-UHFFFAOYSA-L

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (argentometric)99.0-103.0%Barium (Ba)0.05% maxCalcium (Ca)0.05% maxHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.005% maxIron (Fe)5ppm maxMagnesium (Mg)2ppm maxSulfate (SO4)0.001% maxpH of a 5% solution at 25°C5.0-7.0

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service