Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0597

Supelco

Sodium hydroxide solution

50% W/W, Reagent TITRISTAR

Synonym(s):

normality

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352316

Product Name

Sodium Hydroxide Solution 50% W/W, Reagent TITRISTAR

agency

traceable to NIST

Quality Level

vapor pressure

2 hPa ( 20 °C)

pH

14 (20 °C, 89 g/L in H2O)

bp

135 °C/1013 hPa

mp

12 °C

shipped in

ambient

storage temp.

room temp

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service