SX0590
Sodium hydroxide
pellets, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Sodium hydroxide, Caustic Soda,Soda Lye, Caustic Soda, ‘Caustic soda’
About This Item
Recommended Products
Product Name
Sodium Hydroxide Pellets, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
grade
ACS reagent
Quality Level
vapor density
>1 (vs air)
vapor pressure
<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)
assay
99-100% (NaOH)
form
pellets
impurities
≤1% Sodium carbonate
pH
>14 (20 °C, 100 g/L in H2O)
mp
318 °C (lit.)
solubility
1090 g/L
density
2.13 g/cm3 at 20 °C
anion traces
chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.0005%
silicate (as SiO2): ≤0.001%
sulfate (SO42-): ≤0.0005%
cation traces
Al: ≤0.0005%
Ca: ≤0.0005%
Fe: ≤0.0005%
Hg: ≤0.1 ppm
K: ≤0.02%
Mg: ≤0.002%
Ni: ≤0.0005%
heavy metals (as Ag): ≤0.0005%
shipped in
ambient
storage temp.
room temp
SMILES string
[OH-].[Na+]
InChI
1S/Na.H2O/h;1H2/q+1;/p-1
InChI key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Appearance of solution: Passes test
Assay (NaOH): 99-100%
Calcium (Ca): 0.0005% max
Chloride (Cl): 0.0005% max
Heavy metals (as Ag): 0.0005% max
Identity: Passes test
Iron (Fe): 0.0005% max
Magnesium (Mg): 0.002% max
Mercury (Hg): 0.1ppm max
Nickel (Ni): 0.0005% max
Nitrogen compounds (as N): 0.0003% max
Phosphate (PO4): 0.0005% max
Potassium (K): 0.02% max
Silicate (SiO2): 0.001% max
Sodium carbonate: 1.0% max
Sulfate (SO4): 0.0005% max
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service