Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0143E

Supelco

Silica Gel, Desiccant

Grade H Type II, size 10-12 mesh

Synonym(s):

Silica gel, Amorphous Silicon Dioxide, Amorphous Silica

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
EC Index Number:
231-545-4

form

crystals

size

10-12 mesh

particle size

6-12 mesh (>=80%)

bp

2230 °C

mp

>1600 °C

shipped in

ambient

storage temp.

room temp

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Grade H Type II 6-12 Mesh

Analysis Note

Color: White
Form: Crystals
Mesh size (6-12): 80% min

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service