Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ST1682

Sigma-Aldrich

Anti-COPS2 Mouse mAb (4B12)

liquid, clone 4B12, Calbiochem®

Synonym(s):

Anti-COP9 Singlasome Complex Subunit 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

4B12, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... COPS2(9318)

General description

Purified mouse monoclonal antibody. Recognizes the ~50 kDa COPS2 protein.
Recognizes the ~50 kDa COPS2 protein in PC-12, HeLe NE,and Raw 264.7 cells.
This Anti-COPS2 Mouse mAb (4B12) is validated for use in Immunoblotting for the detection of COPS2.

Immunogen

A recombinant polypeptide corresponding to amino acids of 344-444 human COPS2, expressed as a GST fusion protein

Application




Immunoblotting (1-5 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
PC-12 cells, HeLe NE cells, Raw 264.7 cells

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service