ST1600
Anti-ADAM12 Mouse mAb (1G3)
liquid, clone 1G3, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
1G3, monoclonal
form
liquid
does not contain
preservative
species reactivity
human
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG3
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... ADAM12(8038)
General description
Anti-ADAM12, mouse monoclonal, clone 1G3, recognizes ADAM12 protein in human. It is validated for ELISA and Western blotting.
Purified mouse monoclonal antibody. Recognizes the ADAM12 protein.
Recognizes the ADAM12 protein.
Immunogen
A recombinant polypeptide corresponding to amino acids 208-305 of human ADAM12, expressed as a GST fusion protein
Application

ELISA (see comments)
Immunoblotting (1-5 g/ml)
Warning
Toxicity: Regulatory Review (Z)
Physical form
In PBS, pH 7.2.
Reconstitution
Following initial thaw, aliquot and freeze (-20°C).
Other Notes
For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.
Zhou, Z., et al. 2008. Clin. Exp. Metastasis.25 537.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service