ST1203
Anti-PGC-1α Mouse mAb (1F3.9)
liquid, clone 1F3.9, Calbiochem®
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
1F3.9, monoclonal
form
liquid
does not contain
preservative
species reactivity
human, rat, mouse
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG
shipped in
wet ice
storage temp.
−70°C
target post-translational modification
unmodified
Gene Information
human ... PPARGC1A(10891)
General description
Immunogen
Application



Immunoblotting (1 g/ml)
Immunocytochemistry (5-10 g/ml)
Immunohistochemistry (5-10 g/ml)
Immunoprecipitation (1:100)
Warning
Physical form
Analysis Note
Brown adipose tissue from mice exposed to cold
Other Notes
Zhang, Y. et al. 2009. J. Biol. Chem.284, 32813.
Lai, L., et al. 2008. Genes Dev.14, 1948.
Rodgers, J.T., et al. 2008. FEBS Lett.582, 46.
Mazzucotelli, A., et al. 2007. Diabetes10, 2467.
Nemoto, S., et al. 2005. J. Biol. Chem.16, 16456.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service