ST1029
PhosphoDetect Anti-Fos (pSer³⁷⁴) Mouse mAb (34E4)
lyophilized, clone 34E4, Calbiochem®
Select a Size
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
34E4, monoclonal
form
lyophilized
contains
≤0.1% sodium azide as preservative (antibody only)
species reactivity
human
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG1
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pSer374)
General description
Immunogen
Application

ELISA (0.1 g/ml)
Immunoblotting (0.5 g/ml, chemiluminescence)
Warning
Physical form
Reconstitution
Analysis Note
HepG2 cells treated with EGF
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service