SIMPAK0D3
SimPak® Purification Cartridge
For use with Simplicity® systems with DI containing high silt water feed
Synonym(s):
SimPak® 3 Purification Cartridge
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104212
Recommended Products
description
For Simplicity® systems with DI containg high silt water feed
packaging
pkg of 1 unit
compatibility
for use with Simplicity® UV
for use with Simplicity®
Looking for similar products? Visit Product Comparison Guide
Application
SimPak® 3 Purification Cartridge for use with Simplicity® systems with DI containing high silt water feed
Legal Information
SIMPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service