Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCT074

Sigma-Aldrich

BioTracker fVF2 Voltage Sensitive Dye

Voltage-sensitive fluorescent dye for live cell imaging of membrane potential dynamics in excitable cells like neurons and cardiomyocytes.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

form

lyophilized (red)

technique(s)

cell based assay: suitable

detection method

fluorometric

General description

Optical methods for measuring biochemical and biophysical events in living cells provide a powerful approach for monitoring cellular physiology in a non-invasive and high-throughput manner. Changes in the electrochemical potential across the plasma membrane of cells profoundly shapes cellular physiology. Excitable cells, such as neurons and cardiomyocytes, employ a consortium of ion channels both to maintain tight control over their membrane potential (Vm) and to initiate and propagate rapid changes in Vm. Rapid changes in Vm such as an action potential (AP) drive the unique physiology of these excitable cells. Disruption of the frequency, timing, and/or shape of APs is linked to serious human diseases ranging from epilepsy to long QT syndrome. The gold standard for measuring Vm and APs in live cells is electrophysiology: which is highly invasive, low-throughput, and difficult to interpret. Optical recording of the membrane potential using voltage-sensitive fluorescent indicators provides an attractive alternative to probing Vm and AP dynamics in multiple cells, in monolayers, or three-dimensional tissue.BioTracker fVF2 Voltage Sensitive Dye readily detects fast changes in membrane potential in mammalian neurons and cardiomyocytes. The BioTracker dye combines a xanthene-based dye as a fluorescent reporter and a conjugated molecular wire that localizes the indicator to the cell membrane and facilitates photo-induced electron transfer (PeT) from an electron-rich aniline donor to the fluorophore within the low-dielectric environment of the lipid bilayer. The BioTracker dye displays a substantially reduced phototoxicity in cardiomyocytes relative to that of VF2.1.Cl, allowing for prolonged, continuous measurement of cardiomyocyte activity. The dye also exhibits adequate sensitivity and excellent brightness in cells for reporting AP waveforms in neurons and cardiomyocytes with high SNRs.Spectral Properties:Absorption: 520 nmEmission: 535 nmResearch Category:Cardiovascular DiseaseDevelopmental NeuroscienceLive Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service