Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCT067

BioTracker IRI-1 b-Amyloid Plaque Live Cell Dye

Live cell imaging probe for beta-amyloid plaques to study neurodegenrative diseases such as Alzheimer′s disease.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
Pricing and availability is not currently available.

form

lyophilized

detection method

fluorometric

General description

Alzheimer′s disease (AD) is a common progressiveneurodegenerative disease eventually leading to fatality.Significant accumulation of pathological proteinaceous depositsin the brain, including Aβ plaques, are present decades beforesymptoms can be perceived. As such, the delay betweenpathological brain chemistry changes and symptoms creates awindow of opportunity for potential treatments.BioTracker IRI-1 b-Amyloid plaque live cell dye is a near-infrared-emissive two-photon microscopy (TPM) dye with high affinity for Aß-fibrils. The dye demonstrates a ~167-fold increase in fluorescence in the presence of Aß-fibrils. SCT067 achieves this while maintaining large two-photon cross-section, a large signal to background ratio, high BBB permeability, and emission reaching into the near-infrared (NIR, 650 nm). This dye has been used for ex-vivo and in vivo TPM imaging of Aß-fibrils in mouse brains. Spectral Properties:For Two-photon microscopy.Excitation: 850 nmEmission: 580- 779 nmResearch Categories:Alzheimer′s DiseaseAutoimmune diseaseLive Cell DyeNeurodegenerative DiseasesNeurofilament & Neuron MetabolismNeuroscience

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service