SCHVU05RE
Stericup-HV Sterile Vacuum Filtration System
500 mL process volume, 0.45 µm pore size, PVDF membrane, pkg of 12, radio-sterilized
About This Item
Recommended Products
material
PVDF membrane
polystyrene
sterility
sterile
feature
holdup volume 2.4 mL
hydrophilic
manufacturer/tradename
Stericup®
parameter
45 °C max. temp.
500 mL process volume
500 mL sample volume
technique(s)
sterile filtration: suitable
H
263 mm
diam.
12.5 cm
filter diam.
73 mm
filtration area
40 cm2
funnel capacity
500 mL
matrix
Durapore®
pore size
0.45 μm nominal pore size
0.45 μm pore size
fitting
inlet funnel
outlet flask (Receiver)
shipped in
ambient
General description
Features & Benefits
Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications
Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component
Application
Principle
Physical form
Preparation Note
Gamma irradiation
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service