SCGPT02RE
Steritop Threaded Bottle Top Filter
Sterile, 250 mL process volume, 0.22 µm pore size, GP, 45 mm, polyethersulfone membrane, pkg of 12
About This Item
Recommended Products
material
polyethersulfone membrane
polystyrene
Quality Level
sterility
sterile
feature
holdup volume 2.4 mL
hydrophilic
manufacturer/tradename
Steritop®
parameter
250 mL process volume
45 °C max. temp.
application(s)
sterile filtration: suitable
H
95 mm
diam.
12.5 cm
filter diam.
45 mm
filtration area
40 cm2
funnel capacity
250 mL
matrix
Express PLUS™
pore size
0.22 μm nominal pore size
0.22 μm pore size
test parameters
sample volume: 250 mL
fitting
inlet funnel
45 mm outlet thread(s)
shipped in
ambient
General description
Features & Benefits
Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications
Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service